Learn More
Desthiazolylmethyloxycarbonyl Ritonavir-d6, TRC
Product Code. 30356071
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356071 10 mg 10mg
30355052 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30356071 Supplier Toronto Research Chemicals Supplier No. TRCD29743210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1217772-02-5
Molecular Formula C32 D6 H39 N5 O3 S
Molecular Weight (g/mol) 585.83
Synonym (2S)-N-[(1S,3S,4S)-4-Amino-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]-3-methyl-2-[[[methyl[[2-(1-methylethyl-d6)-4-thiazolyl]methyl]amino]carbonyl]amino]butanamide
IUPAC Name (2S)-N-[(1S,3S,4S)-4-amino-1-benzyl-3-hydroxy-5-phenyl-pentyl]-3-methyl-2-[[methyl-[[2-[2,2,2-trideuterio-1-(trideuteriomethyl)ethyl]thiazol-4-yl]methyl]carbamoyl]amino]butanamide
SMILES [2H]C([2H])([2H])C(c1nc(CN(C)C(=O)N[C@@H](C(C)C)C(=O)N[C@H](C[C@H](O)[C@@H](N)Cc2ccccc2)Cc3ccccc3)cs1)C([2H])([2H])[2H]
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C32H45N5O3S/c1-21(2)29(36-32(40)37(5)19-26-20-41-31(35-26)22(3)4)30(39)34-25(16-23-12-8-6-9-13-23)18-28(38)27(33)17-24-14-10-7-11-15-24/h6-15,20-22,25,27-29,38H,16-19,33H2,1-5H3,(H,34,39)(H,36,40)/t25-,27-,28-,29-/m0/s1/i3D3,4D3
Formula Weight 585.362
Chemical Name or Material Desthiazolylmethyloxycarbonyl Ritonavir thiazolyl-isopropyl-1,1,1,3,3,3-d6