missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Destrin Antibody (OTI2F7), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71669APCCY7
This item is not returnable.
View return policy
Description
Destrin Monoclonal specifically detects Destrin in Human, Mouse, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.
Specifications
| Destrin | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| DSTN | |
| Full length recombinant protein of human Destrin (NP_006861) produced in HEK293T cells. | |
| 0.1 mL | |
| 11034.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI2F7 | |
| Flow Cytometry | |
| ACTDPdestrin, Actin-depolymerizing factor, ADFbA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)), bA462D18.2, destrin (actin depolymerizing factor), DSN | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Canine, Monkey, Primate | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction