missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dexamethasone Monoclonal Antibody (6D7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549708
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Specifications
| Dexamethasone | |
| Monoclonal | |
| 4.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of DEX hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 6D7 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction