missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt (400,000-600,000 M.Wt), MP Biomedicals™

Product Code. 11474352 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
10 g
50 g
100 g
Unit Size:
100g
10g
1g
50g
This item is not returnable. View return policy

Product Code. 11474352

Brand: MP Biomedicals 0216011110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Color White
Physical Form Powder
Quantity 10 g
IUPAC Name ethyl 4-aminobenzoate
Formula Weight 400,000 to 600,000
Chemical Name or Material Dextran Sulfate Sodium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.