missing translation for 'onlineSavingsMsg'
Learn More

Dextro Mepromazine Maleate, TRC

Product Code. 30350424
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350424 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350424 Supplier Toronto Research Chemicals Supplier No. TRCD29925025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dextro Mepromazine Maleate
CAS 2622-31-3
Molecular Formula C19 H24 N2 O S . C4 H4 O4
Quantity 25 mg
Synonym (betaS)-2-Methoxy-N,N,beta-trimethyl-10H-phenothiazine-10-propanamine, (+)-10-[3-(Dimethylamino)-2-methylpropyl]-2-methoxyphenothiazine, (+)-Methotrimeprazine, Dextromepromazine, d-Methotrimeprazine
InChI Formula InChI=1S/C19H24N2OS.C4H4O4/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)23-19-10-9-15(22-4)11-17(19)21;5-3(6)1-2-4(7)8/h5-11,14H,12-13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-;/m0./s1
SMILES COc1ccc2Sc3ccccc3N(C[C@@H](C)CN(C)C)c2c1.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;(2S)-3-(2-methoxyphenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine
Molecular Weight (g/mol) 444.544
Formula Weight 444.172
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.