missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAST205 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-22986JF549
This item is not returnable.
View return policy
Description
MAST205 Polyclonal antibody specifically detects MAST205 in Human samples. It is validated for Western Blot
Specifications
| MAST205 | |
| Polyclonal | |
| Western Blot | |
| EC 2.7.11, EC 2.7.11.1, FLJ39200, KIAA0807RP4-533D7.1, MAST205microtubule associated testis specific serine/threonine protein kinase, microtubule associated serine/threonine kinase 2, microtubule-associated serine/threonine-protein kinase 2, MTSSK | |
| The immunogen recognized by this antibody maps to a region between residue 1748 and 1798 of human microtubule associated serine/threonine kinase 2 using the numbering given in entry NP_055927.2 (GeneID 23139). | |
| 0.1 mL | |
| Protein Kinase | |
| 23139 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction