Learn More
Tocris Bioscience™ DFHBI
GREENER_CHOICE
Mimic of GFP fluorophore for imaging RNA in living cells
Brand:  Tocris Bioscience™ 5609/50
Encompass
Product Code. 15726976
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
DFHBI is a mimic of green fluorescent protein (GFP) fluorophore for imaging RNA in living cells. DFHBI fluorescence is activated by binding to Spinach2 or Broccoli aptamers. Exhibits peak excitation maxima of 447 nm and peak fluorescence emission of 501 nm when bound to Spinach2.
Specifications
Specifications
(5Z)-5-[(3,5-Difluoro-4-hydroxyphenyl)methylene]-3,5-dihydro-2,3-dimethyl-4H-imidazol-4-one
1241390-29-3