Learn More
Tocris Bioscience™ DG 041
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
DG 041 is a potent and selective EP3 antagonist (IC50 = 4.6 nM). Inhibits PGE2 facilitation of platelet aggregation in vitro and ex vivo. Displays good plasma and metabolic stability. Does not affect bleeding time in rats. Antiplatelet and antithrombotic.
Specifications
Specifications
| Chemical Name or Material | (2E)-3-[1-[(2,4-Dichlorophenyl)methyl]-5-fluoro-3-methyl-1H-indol-7-yl]-N-[(4,5-dichloro-2-thienyl)sulfonyl]-2-propenamide |
| CAS | 861238-35-9 |
| Quantity | 50 mg |
| Target | Prostanoid Receptor Antagonists |
| Molecular Formula | C23H15Cl4FN2O3S2 |
| Purity | 0.98 |