missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NAP-2/PPBP ELISA Development Kit (ABTS), PeproTech®
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
Description
Members of the a-chemokine subfamily of inducible, secreted, pro-inflammatory cytokines contain a similar motif, in which the first two cysteine residues are separated by a single residue (Cys-X-Cys), and are also chemotactic for neutrophils. The platelet basic protein (PBP), a member of the a-chemokine family, resides in the a-granules of platelets and is released upon their activation. Proteolytic cleavage of the amino terminus of PBP leads to the generation of several peptides, which include mature PBP, connective tissue-activating peptide III (CTAP III, also designated low affinity platelet factor IV (LA-PF4)), b-thromboglobulin (b-TG), and neutrophil-activating peptide 2 (NAP-2). PBP and its N-truncated derivatives mediate inflammation and wound healing. Specifically, NAP-2 activates chemotaxis and degranulation in neutrophils during inflammation. The gene encoding human PBP maps to chromosome 4q12-q13.
Specifications
Specifications
| Accession Number | P02775 |
| Assay Range | Lot-specific |
| Assay Sensitivity | Lot-specific |
| Conjugate | HRP |
| Product Type | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 5473 |
| Gene Symbol | PPBP |
| Kit Contents | Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate |
| Label or Dye | HRP |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction