missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGAT2, Goat anti-Human, Polyclonal Antibody, Abnova™
Goat Polyclonal Antibody
Brand: Abnova PAB7220.100ug
This item is not returnable.
View return policy
Description
Sequence: C-KLEHPTQQDIDLYHSpecifications
| DGAT2 | |
| Polyclonal | |
| Unconjugated | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| DKFZp686A15125/HMFN1045 | |
| Goat | |
| 100 μg | |
| Primary | |
| This antibody is not expected to cross-react with other DGAT members. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| ELISA (1:128000) Sandwhich ELISA (0.5-1 ug/mL) Western blot (0.3-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3 ug/mL) The optimal working dilution should be determined by the end user. | |
| diacylglycerol O-acyltransferase homolog 2 (mouse) | |
| DGAT2 | |
| A synthetic peptide corresponding to near C terminus of human DGAT2. | |
| RUO | |
| 84649 | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction