Learn More
Invitrogen™ DGAT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5144451
Product Code. 17979912
  • € 378.00 / 100µL
Description
DGAT3 Polyclonal Antibody for Western Blot, ELISA
Involved in triacylglycerol (TAG) biosynthesis (PubMed:22760209, PubMed:30467384). Catalyzes the acylation of the sn-3 hydroxy group of sn-1,2-diacylglycerol using acyl-CoA (PubMed:22760209, PubMed:30467384). May preferentially use linolenoyl-CoA as substrate and to a lesser extent linoleoyl-CoA (PubMed:22760209). May contribute to the active recycling of linoleate and linolenate into TAG when seed oil breakdown is blocked (PubMed:22760209).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DGAT3
Polyclonal
Unconjugated
DGAT3
cytosolic DGAT3 At1g48300 F11A17.15; Diacylglycerol O-acyltransferase 3
Rabbit
Protein A/G
RUO
841250
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot
4.5 mg/mL
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Q9C5W0
DGAT3
Recombinant Arabidopsis thaliana DGAT3 protein.
100 μL
Primary
Plant