missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGCR14 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36530DL594
This item is not returnable.
View return policy
Description
DGCR14 Polyclonal antibody specifically detects DGCR14 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DGCR14 | |
| Polyclonal | |
| Rabbit | |
| 0.1 mL | |
| Store at 4°C in the dark. |
| Western Blot | |
| DyLight 594 | |
| The immunogen recognized by the antibody maps to a region between residue 426 to 476 of human DiGeorge Syndrome Critical Region Gene 14 using the numbering given in entry NP_073210.1. | |
| RUO |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction