Learn More
Invitrogen™ DGCR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113825
Description
Immunogen sequence: AESCYEKSSF LCKRSQTCVD IKDNVVDEGF YFTPKGDDPC LSCTCHGGEP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Deletions of the 22q11.2 have been associated with a wide range of developmental defects (notably DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome and isolated conotruncal cardiac defects) classified under the acronym CATCH 22. The DGCR2 gene encodes a novel putative adhesion receptor protein, which could play a role in neural crest cells migration, a process which has been proposed to be altered in DiGeorge syndrome.
Specifications
| DGCR2 | |
| Polyclonal | |
| Unconjugated | |
| DGCR2 | |
| 9930034O06Rik; Dgcr2; Dgsc; DGS-C; DiGeorge syndrome critical region gene 2; DiGeorge syndrome critical region protein 2; DiGeorge syndrome gene c; DiGeorge syndrome protein C; IDD; integral membrane protein deleted in DiGeorge syndrome; integral membrane protein DGCR2/IDD; KIAA0163; Lan; mKIAA0163; seizure-related membrane-bound adhesion protein; Sez12; Sez-12 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 360742, 9993 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P98153 | |
| DGCR2 | |
| Synthetic peptide directed towards the middle region of human DGCR2. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.