Learn More
DGCR8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-21407-25ul
 View more versions of this product
Product Code. 18649594
  • ONLINE EXCLUSIVE PRICE
    € 450.00
    € 425.25 / 25µL
    Save € 24.75
    5% Off
Description
DGCR8 Polyclonal antibody specifically detects DGCR8 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DGCR8
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
C22orf12, DGCRK6chromosome 22 open reading frame 12, DiGeorge syndrome critical region 8, DiGeorge syndrome critical region gene 8, Gy1, microprocessor complex subunit DGCR8, pasha
This antibody was developed against Recombinant Protein corresponding to amino acids: IDGVTYGSGTASSKKLAKNKAARATLEILIPDFVKQTSEEKPKDSEELEYFNHISIEDSRVYELTSKAGLLSPYQILHECLKRNHGMGDTSIKFEVVPG
25 μg
Epigenetics, Stem Cell Markers
54487
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol
Rabbit
Affinity purified