missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DGK-beta Polyclonal antibody specifically detects DGK-beta in Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | DGK-beta |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | 90 kDa diacylglycerol kinase, DAG kinase beta, DAGK2, DGK-beta, diacylglycerol kinase beta, diacylglycerol kinase, beta (90kD), diacylglycerol kinase, beta 90kDa, Diglyceride kinase beta, EC 2.7.1, EC 2.7.1.107, KIAA0718DGKDGK-BETA |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 544-773 of human DGKB (NP_663733.1). LMKILKDIENSTEIMLDRWKFEVIPNDKDEKGDPVPYSIINNYFSIGVDASIAHRFHIMREKHPEKFNSRMKNKFWYFEFGTSETFSATCKKLHESVEIECDGVQIDLINISLEGIAILNIPSMHGGSNLWGESKKRRSHRRIEKKGSDKRTTVTDAKELKFASQDLSDQLLEVVGLEGAMEMGQIYTGLKSAGRRLAQCSCVVIRTSKSLPMQIDGEPWMQTPCTVSTE |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?