missing translation for 'onlineSavingsMsg'
Learn More

DGK-beta Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. p-200062126 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18697440 0.02 mL 0.02mL
18673160 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18697440 Supplier Novus Biologicals Supplier No. NBP2920280.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DGK-beta Polyclonal antibody specifically detects DGK-beta in Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGK-beta
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 90 kDa diacylglycerol kinase, DAG kinase beta, DAGK2, DGK-beta, diacylglycerol kinase beta, diacylglycerol kinase, beta (90kD), diacylglycerol kinase, beta 90kDa, Diglyceride kinase beta, EC 2.7.1, EC 2.7.1.107, KIAA0718DGKDGK-BETA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 544-773 of human DGKB (NP_663733.1). LMKILKDIENSTEIMLDRWKFEVIPNDKDEKGDPVPYSIINNYFSIGVDASIAHRFHIMREKHPEKFNSRMKNKFWYFEFGTSETFSATCKKLHESVEIECDGVQIDLINISLEGIAILNIPSMHGGSNLWGESKKRRSHRRIEKKGSDKRTTVTDAKELKFASQDLSDQLLEVVGLEGAMEMGQIYTGLKSAGRRLAQCSCVVIRTSKSLPMQIDGEPWMQTPCTVSTE
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1607
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.