missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGK-theta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69208
This item is not returnable.
View return policy
Description
DGK-theta Polyclonal antibody specifically detects DGK-theta in Human samples. It is validated for Western Blot
Specifications
| DGK-theta | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| DAG kinase theta, DAGK, DAGK4EC 2.7.1.107, DAGK7, DGK-theta, diacylglycerol kinase theta, diacylglycerol kinase, theta (110kD), diacylglycerol kinase, theta 110kDa, Diglyceride kinase theta | |
| Synthetic peptides corresponding to DGKQ (diacylglycerol kinase, theta 110kDa) The peptide sequence was selected form the middle region of DGKQ. Peptide sequence DAELSLDFHQAREEEPGKFTSRLHNKGVYVRVGLQKISHSRSLHKQIRLQ. The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μg | |
| Lipid and Metabolism | |
| 1609 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1.0 μg/mL | |
| P52824 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction