Learn More
DGKA Monoclonal Antibody (OTI4H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504069
Encompass_Preferred
 View more versions of this product
Product Code. 17590722
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It acts as a modulator that competes with protein kinase C for the second messenger diacylglycerol in intracellular signaling pathways. It also plays an important role in the resynthesis of phosphatidylinositols and phosphorylating diacylglycerol to phosphatidic acid. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified.
Specifications
Specifications
DGKA
Monoclonal
0.62 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P23743
DGKA
Full length human recombit protein of human DGKA produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI4H10
Unconjugated
DGKA
DAGK, DAGK1, DGK-alpha
Mouse
Affinity Chromatography
RUO
1606
-20° C, Avoid Freeze/Thaw Cycles