missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGKB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24320-1-AP-150UL
This item is not returnable.
View return policy
Description
Diacylglycerol (DAG) is an allosteric activator of protein kinase C. DAG also participates in regulating RAS and RHO family proteins by activating the guanine nucleotide exchange factors VAV and RASGRP1. DAG is also involved in the synthesis of phospholipids and triacylglycerols. Tight regulation of DAG levels is achieved via DAG kinases (DGKs), which remove DAG by phosphorylate it to phosphatidic acid. Several mammalian isozymes of DAGK have been identified.Specifications
| DGKB | |
| Polyclonal | |
| Unconjugated | |
| DGKB | |
| 90 kDa diacylglycerol kinase, DAG kinase beta, DAGK2, DGK, DGK BETA, DGKB, Diacylglycerol kinase beta, Diglyceride kinase beta, KIAA0718 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1607 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y6T7 | |
| DGKB | |
| DGKB Fusion Protein Ag19809 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction