missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DGKD Polyclonal Antibody

Product Code. 17158863
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17158863 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17158863 Supplier Invitrogen™ Supplier No. PA5110941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TQMDQFGQAA GVLIHSIREI AQSHRDMEQE LAHAVNASSK SMDRVYGKPR TTEGLNCSFV LEMVNNFRAL RSETELLLSG KMALQLDPPQ KEQLGSALAE MDRQLRRLAD TPWLCQSAEP GDEESVMLDL A Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 84%.

Diacylglycerol kinases (DGKs) phosphorylate diacylglycerol (DAG) to produce phosphatidic acid. DAG and phosphatidic acid are lipids that act as second messengers in signaling cascades. DGK-alpha influences cell activation and secretion of lethal exosomes, which in turn control cell death. DGK-beta is abundant in restricted brain regions such as the caudate putamen and olfactory tubercle. DGK-gamma encodes full-length and truncated transcripts that are present in a range of human tissues, with greatest expression observed in retina. DGK-delta is most abundant in skeletal muscle. DGK-epsilon shows specificity for arachidonylcontaining diacylglycerol and is expressed predominantly in testis. DGK-theta is most abundant in the cerebellum and hippocampus. DGK-iota is present in brain and retina as a predominant transcript of more than 12 kb, including a long 3-prime untranslated region, with additional low abundance transcripts of 9.5 and 7.5 kb. DGK-eta is closely related to DGK-delta. DGK-zeta is most abundant in brain and muscle. DGKs have structural motifs that play regulatory roles, and these motifs form the basis for dividing the DGKs into five subtypes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGKD
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DGKD
Gene Accession No. Q16760
Gene Alias 130 kDa diacylglycerol kinase; AI841987; D330025K09; DAG kinase delta; DGK delta; DGKD; dgkd-2; DGKdelta; DGK-delta; Diacylglycerol kinase delta; diacylglycerol kinase, delta; diacylglycerol kinase, delta 130kDa; Diglyceride kinase; diglyceride kinase delta; EC 2.7.1.107; FLJ26930; hypothetical protein LOC513389; KIAA0145; RGD1563309
Gene Symbols DGKD
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human DGKD. Recombinant protein control fragment (Product #RP-104581).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8527
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.