missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DGKE Polyclonal Antibody
GREENER_CHOICE

Product Code. 15791491 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15791491 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15791491 Supplier Invitrogen™ Supplier No. PA553558

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RDTDLFSQPT YCCVCAQHIL QGAFCDCCGL RVDEGCLRKA DKRFQCKEIM LKNDTKVLDA MPHHWIRGNV PLCSYCMVCK QQCGCQPKLC DYRCIWCQKT VHDECMKNSL KNEKCDFGEF KNLIIPPSYL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

Diacylglycerol (DAG) influences numerous cell signaling cascades by functioning as an intracellular, allosteric activator of protein kinase C (PKC), and as a potent activator of guanine nucleotide exchange factors. In order to maintain cellular homeostasis, intracellular DAG levels are tightly regulated by diacylglycerol kinases (DGKs, DAGKs), which phosphorylate DAG to phosphatidic acid, thus removing DAG. Human DGK-alpha (80 kDa), -beta (90 kDa), and - gamma (90 kDa) have calcium-binding EF-hand motifs at their N termini and are classified as type I DGKs. Human DGK- delta (130 kDa) and DGK-i (130 kDa) contain N-terminal pleckstrin homology (PH) domains and are classified as type II. Human DGK- epsilon (64 kDa) contains no identifiable regulatory domains and is classified as a type III DGK. Human DGK- zeta (104 kDa) and -iota (130 kDa) possess C-terminal ankyrin repeats and are classified as type IV DGKs. Human DGK-theta (110 kDa) contains 3 cysteine-rich domains and a PH domain and is classified as a type V DGK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGKE
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dgke
Gene Accession No. P52429
Gene Alias AHUS7; C87606; DAG kinase epsilon; DAGK5; DAGK6; DGK; DGKE; DGK-epsilon; Diacylglycerol kinase epsilon; diacylglycerol kinase, epsilon; diacylglycerol kinase, epsilon 64kDa; diglyceride kinase epsilon; EC 2.7.1.107; NPHS7
Gene Symbols Dgke
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DGKE. Recombinant protein control fragment (Product #RP-90372).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8526
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.