missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHCR7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577942
This item is not returnable.
View return policy
Description
Positive Control: HepG2 (boiled), HepG2 MF (boiled), HepG2 (unboiled), HepG2 MF (unboiled) Predicted Reactivity: Bovine (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Production of cholesterol by reduction of C7-C8 double bond of 7-dehydrocholesterol (7-DHC).
Specifications
| DHCR7 | |
| Polyclonal | |
| Unconjugated | |
| DHCR7 | |
| 7-dehydrocholesterol reductase; 7-DHC reductase; AA409147; D7SR; delta-7-dehydrocholesterol reductase; Delta7-sterol reductase; Dhcr7; putative sterol reductase SR-2; SLOS; Sterol Delta(7)-reductase; sterol delta-7-reductase; Sterol reductase SR-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13360, 1717, 64191 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.46 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O88455, Q9UBM7, Q9Z2Z8 | |
| DHCR7 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human DHCR7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction