missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHCR7 Recombinant Rabbit Monoclonal Antibody (HL2764)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556678
This item is not returnable.
View return policy
Description
DHCR7 Recombinant Monoclonal Antibody for Western Blot
Production of cholesterol by reduction of C7-C8 double bond of 7-dehydrocholesterol (7-DHC).
Specifications
| DHCR7 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9UBM7 | |
| DHCR7 | |
| Recombinant fragment of human DHCR7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2764 | |
| Unconjugated | |
| DHCR7 | |
| 7-dehydrocholesterol reductase; 7-DHC reductase; AA409147; D7SR; delta-7-dehydrocholesterol reductase; Delta7-sterol reductase; Dhcr7; putative sterol reductase SR-2; SLOS; Sterol Delta(7)-reductase; sterol delta-7-reductase; Sterol reductase SR-2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1717 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction