missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHFR Mouse anti-Canine, Human, Clone: OTI6G7, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500543
This item is not returnable.
View return policy
Description
Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, and is required for the synthesis of purines, thymidylic acid and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.Specifications
| DHFR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P00374 | |
| DHFR | |
| Full-length protein expressed in 293T cell transfected with human DHFR expression vector | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6G7 | |
| Unconjugated | |
| DHFR | |
| DHFRP1, DYR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1719, 479165 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction