missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHFR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA514267
This item is not returnable.
View return policy
Description
DHFR Polyclonal Antibody for Western Blot, IHC (P)
Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, and is required for the synthesis of purines, thymidylic acid and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.
Specifications
| DHFR | |
| Polyclonal | |
| Unconjugated | |
| DHFR | |
| 8430436I03Rik; AA607882; AI662710; AW555094; Dhfr; Dhfr1; DHFRP1; Dihydrofolate reductase; Dihydrofolate reductase 1 (active); DYR | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 13361, 1719, 24312 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.0 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P00374, P00375, Q920D2 | |
| DHFR | |
| KLH conjugated synthetic peptide between 135-164 amino acids from the C-terminal region of mouse DHFR. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction