missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHFRL1 Antibody (OTI1E2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72459APC
This item is not returnable.
View return policy
Description
DHFRL1 Monoclonal antibody specifically detects DHFRL1 in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| DHFRL1 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 200895 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| OTI1E2 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence | |
| DHFRP4, dihydrofolate reductase pseudogene 4, dihydrofolate reductase-like 1, dihydrofolate reductase-like protein 1, EC 1.5.1.-, EC 1.5.1.3, FLJ16119 | |
| Human recombinant protein fragment corresponding to amino acids 4-185 of human DHFRL1(NP_789785) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction