missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHFRL1 Monoclonal Antibody (OTI1E2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525633
This item is not returnable.
View return policy
Description
DHFRL1 Monoclonal Antibody for Western Blot, ICC/IF
Key enzyme in folate metabolism. Contributes to the de novo mitochondrial thymidylate biosynthesis pathway. Required to prevent uracil accumulation in mtDNA. Binds its own mRNA and that of DHFR.
Specifications
| DHFRL1 | |
| Monoclonal | |
| 0.73 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86XF0 | |
| Dhfr-rs1, DHFR2 | |
| Human recombinant protein fragment corresponding to amino acids 4-185 of DHFRL1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Canine, Monkey | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| OTI1E2 | |
| Unconjugated | |
| DHFR2 | |
| DHFR2; DHFRL1; DHFRP4; dihydrofolate reductase 2; Dihydrofolate reductase 2, mitochondrial; dihydrofolate reductase like 1; dihydrofolate reductase pseudogene 4; dihydrofolate reductase, mitochondrial; dihydrofolate reductase-like 1; dihydrofolate reductase-like protein 1; LOW QUALITY PROTEIN: dihydrofolate reductase 2, mitochondrial | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 111208, 200895 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction