missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHODH Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA589141

Product Code. 16557151

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EIGSVTPKPQ EGNPRPRVFR LPEDQAVINR YGFNSHGLSV VEHRLRARQQ KQAKLTEDGL PLGVNLGKNK TSVDAAEDYA EGVRVLGPLA DYLVVNVSSP NTAGLRSLQG KAELRRLLTK VLQERDGLRR VHRPAVLVKI APDLTSQDKE DIASVVKELG IDGLIVTNTT VSRPAGLQGA LRSETGGLSG KPLRDLSTQT IREMYALTQG RVPIIGVGGV SSGQDALEKI RAGASLVQLY TALTFWGPPV VGKVKRELEA LLKEQGFGGV TDAIGADHRR; Positive Samples: HL-60, SW620, HepG2, Mouse heart, Mouse spleen, Mouse kidney, Rat heart, Rat kidney; Cellular Location: Mitochondrion inner membrane, Single-pass membrane protein.

The protein encoded by this gene catalyzes the fourth enzymatic step, the ubiquinone-mediated oxidation of dihydroorotate to orotate, in de novo pyrimidine biosynthesis. This protein is a mitochondrial protein located on the outer surface of the inner mitochondrial membrane. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DHODH
Polyclonal
Unconjugated
DHODH
2810417D19Rik; AI834883; DHOdehase; DHODH; dihydroorotate dehydrogenase; dihydroorotate dehydrogenase (quinone); dihydroorotate dehydrogenase (quinone), mitochondrial; dihydroorotate dehydrogenase, mitochondrial; dihydroorotate oxidase; human complement of yeast URA1; POADS; URA1
Rabbit
Affinity Chromatography
RUO
1723, 56749, 65156
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.82 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35435, Q02127, Q63707
DHODH
Recombinant fusion protein containing a sequence corresponding to amino acids 116-395 of human DHODH (NP_0013522).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.