missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHRS11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569635
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
DHRS11 gene ontology annotations related to this gene include oxidoreductase activity.
Specifications
| DHRS11 | |
| Polyclonal | |
| Unconjugated | |
| DHRS11 | |
| 17-beta-hydroxysteroid dehydrogenase; 3-beta-hydroxysteroid 3-dehydrogenase; ARPG836; dehydrogenase/reductase (SDR family) member 11; dehydrogenase/reductase 11; dehydrogenase/reductase SDR family member 11; DHRS11; Estradiol 17-beta-dehydrogenase; hypothetical protein LOC510264; inactive dehydrogenase/reductase isoform; LOW QUALITY PROTEIN: dehydrogenase/reductase SDR family member 11; RGD1307935; SDR24C1; short chain dehydrogenase/reductase family 24C, member 1; short-chain dehydrogenase/reductase family 24C member 1; spDHRS11; UNQ836/PRO1774 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79154 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6UWP2 | |
| DHRS11 | |
| synthetic peptide directed towards the C terminal region of human DHRS11. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction