missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHRS12 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA571082

Product Code. 15829493

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This target displays homology in the following species: Cow: 86%; Human: 100%; Zebrafish: 77%.

Putative oxidoreductase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DHRS12
Polyclonal
Unconjugated
DHRS12
dehydrogenase/reductase (SDR family) member 12; dehydrogenase/reductase 12; dehydrogenase/reductase SDR family member 12; DHRS12; SDR40C1; short chain dehydrogenase/reductase family 40C, member 1; short-chain dehydrogenase/reductase family 40C member 1; zgc:153679
Rabbit
Affinity chromatography
RUO
79758
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
A0PJE2
DHRS12
synthetic peptide directed towards the C-terminal region of human DHRS12.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.