missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS3 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99196F
This item is not returnable.
View return policy
Description
DHRS3 Polyclonal antibody specifically detects DHRS3 in Human samples. It is validated for Western Blot
Specifications
| DHRS3 | |
| Polyclonal | |
| Western Blot | |
| DD83.1, dehydrogenase/reductase (SDR family) member 3, EC 1.1.1, EC 1.1.1.300, RDH17, Retinal short-chain dehydrogenase/reductase 1, RETSDR1, retSDR1short chain dehydrogenase/reductase family 16C, member 1, Rsdr1, SDR1, SDR16C1, short-chain dehydrogenase/reductase 1, short-chain dehydrogenase/reductase 3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DHRS3. | |
| 0.1 mL | |
| Vision | |
| 9249 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction