missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 463.00
Specifications
| Antigen | DHRS4 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647140
|
Novus Biologicals
NBP2-92009-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18697160
|
Novus Biologicals
NBP2-92009-0.1ml |
0.1 mL |
€ 463.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHRS4 Polyclonal antibody specifically detects DHRS4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DHRS4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 10901 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CR, dehydrogenase/reductase (SDR family) member 4, dehydrogenase/reductase (SDR family) member 4 like 2A, dehydrogenase/reductase SDR family member 4, EC 1.1.1, EC 1.1.1.184, FLJ11008, humNRDR, NADP-dependent retinol dehydrogenase, NRDRNADPH-dependent retinol dehydrogenase/reductase, Peroxisomal short-chain alcohol dehydrogenase, PHCR, PSCD, SCAD-SRLNADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase, SDR25C1, SDR25C2, SDR-SRL, short chain dehydrogenase/reductase family 25C, member 1, short chain dehydrogenase/reductase family 25C, member 2, Short-chain dehydrogenase/reductase family member 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-278 of human DHRS4 (NP_066284.2). MHKAGLLGLCARAWNSVRMASSGMTRRDPLANKVALVTASTDGIGFAIARRLAQDGAHVVVSSRKQQNVDQAVATLQGEGLSVTGTVCHVGKAEDRERLVATAVKLHGGIDILVSNAAVNPFFGSIMDVTEEVWDKTLDINVKAPALMTKAVVPEMEKRGGGSVVIVSSIAAFSPSPGFSPYNVSKTALLGLTKTLAIELAPRNIRVNCLAPGLIKTSFSRMLWMDKEKEESMKETLRIRRLGEPEDCAGIVSFLCSEDASYITGETVVVGGGTPSRL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title