missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS4L2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00317749-B01P
This item is not returnable.
View return policy
Description
DHRS4L2 Polyclonal antibody specifically detects DHRS4L2 in Human samples. It is validated for Western Blot
Specifications
| DHRS4L2 | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAH00663 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| dehydrogenase/reductase (SDR family) member 4 like 2, dehydrogenase/reductase (SDR family) member 4 like 2A3, dehydrogenase/reductase SDR family member 4-like 2, EC 1.1, EC 1.1.1, EC 1.1.1.184, FLJ11525, MGC905, SDR25C3, short chain dehydrogenase/reductase family 25C, member 3 | |
| DHRS4L2 (AAH00663, 1 a.a. - 230 a.a.) full-length human protein. MARLLGLCAWARKSVRLASSRMTRRDPLTNKVALVTASTDGIGFAIARRLAQDRAHVVVSSRKQQNVDQAVATLQGEGLSVTGTVCHVGKAEDRERLVAMAVKLHGGIDILVSNAAVNPFFGSLMDVTEEVWDKTLDINVKAPALMTKAVVPEMEKRGGGSVVIVSSIAAFSPSPGFSPYNVSKTALLGLNNTLAIELAPRNIRVNCLHLDLSRLASAGCSGWTRKKRKA | |
| 0.05 mg | |
| Lipid and Metabolism | |
| 317749 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction