missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS4L2 Monoclonal Antibody (OTI8A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810278S
This item is not returnable.
View return policy
Description
DHRS4L2 Monoclonal antibody specifically detects DHRS4L2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| DHRS4L2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6PKH6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 317749 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8A2 | |
| Unconjugated | |
| DHRS4L2 | |
| dehydrogenase/reductase (SDR family) member 4 like 2; dehydrogenase/reductase (SDR family) member 4 like 2A3; dehydrogenase/reductase 4 like 2; dehydrogenase/reductase SDR family member 4-like 2; DHRS4L2; NADP(H)-dependent retinol dehydrogenase/reductase short isoform-like protein; NADPH-dependent retinol dehydrogenase/reductase-like protein 2; SDR25C3; short chain dehydrogenase/reductase family 25C member 3; short chain dehydrogenase/reductase family 25C, member 3 | |
| Full length human recombinant protein of human DHRS4L2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction