Learn More
DHRS4L2 Monoclonal Antibody (OTI8A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810278
This item is not returnable.
View return policy
Description
DHRS4L2 encodes a member of the short chain dehydrogenase reductase family. The encoded protein may be an NADPH dependent retinol oxidoreductase. Alternate splicing results in multiple transcript variants.Specifications
| DHRS4L2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6PKH6 | |
| DHRS4L2 | |
| Full length human recombit protein of human DHRS4L2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8A2 | |
| Unconjugated | |
| DHRS4L2 | |
| SDR25C3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 317749 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |