missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHRS7C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548632
This item is not returnable.
View return policy
Description
Predicted to react with bovine and mouse based on sequence homology.
Putative oxidoreductase.
Specifications
| DHRS7C | |
| Polyclonal | |
| Unconjugated | |
| DHRS7C | |
| 1110001P11Rik; AI120487; dehydrogenase/reductase (SDR family) member 7C; dehydrogenase/reductase 7C; dehydrogenase/reductase SDR family member 7C; Dhrs7c; RGD1306989; SDR32C2; short chain dehydrogenase/reductase family 32C, member 2; short-chain dehydrogenase/reductase family 32C member 2; SRP-35 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 201140 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A6NNS2 | |
| DHRS7C | |
| KLH conjugated synthetic peptide between 116-145 amino acids from the Central region of human DHRS7C. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction