missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS9 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97017MFV610
This item is not returnable.
View return policy
Description
DHRS9 Polyclonal antibody specifically detects DHRS9 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| DHRS9 | |
| Polyclonal | |
| Western Blot, ELISA, Immunoprecipitation | |
| dehydrogenase/reductase (SDR family) member 9,3alpha-HSD, EC 1.1, EC 1.1.1, EC 1.1.1.105, member 4,3-alpha-HSD, NADP-dependent retinol dehydrogenase/reductase, RDH15, RDH-E2, RDHL3ALPHA-HSD, RDHTBE | |
| Produced in rabbits immunized with purified, recombinant Human DHRS9 (Accession#: NP_005762.2; Arg18-Val319) | |
| 0.1 mL | |
| metabolism | |
| 10170 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction