missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRSX Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97560R
This item is not returnable.
View return policy
Description
DHRSX Polyclonal specifically detects DHRSX in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DHRSX | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CXorf11, dehydrogenase/reductase (SDR family) X chromosome, dehydrogenase/reductase (SDR family) X-linked, DHRS5Xmember 1, EC 1.1, SDR46C1 | |
| Produced in rabbits immunized with E. coli-derived Human DHRSX fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 207063 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction