missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX15 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-586AF488
This item is not returnable.
View return policy
Description
DHX15 Polyclonal antibody specifically detects DHX15 in Human samples. It is validated for Western Blot
Specifications
| DHX15 | |
| Polyclonal | |
| Western Blot | |
| ATP-dependent RNA helicase #46, DBP1DEAH box protein 15, DDX15putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15, DEAH (Asp-Glu-Ala-His) box polypeptide 15, EC 3.6.1, EC 3.6.4.13, HRH2DEAD/H box-15, PRP43, PRPF43, PrPp43p, RNA helicase 2 | |
| The immunogen recognized by this antibody maps to a region between residue 750 and the C-terminus (residue 795) of human DEAH box polypeptide 15 using the numbering given in Swiss-Prot entry O43143 (GeneID 1665). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 1665 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction