missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16161
This item is not returnable.
View return policy
Description
DHX16 Polyclonal specifically detects DHX16 in Human samples. It is validated for Western Blot.
Specifications
| DHX16 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| ATP-dependent RNA helicase #3, DBP2PRO2014, DDX16DEAD/H box 16, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 16, DEAH (Asp-Glu-Ala-His) box polypeptide 16, DEAH-box protein 16, EC 3.6.1, EC 3.6.4.13, KIAA0577, Prp2, PRP8, PRPF2, putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16, RNA helicase | |
| Rabbit | |
| 119 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8449 | |
| Human | |
| IgG |
| Western Blot | |
| 0.52mg/mL | |
| Western Blot 1:500-1:3000 | |
| O60231 | |
| DHX16 | |
| Recombinant protein encompassing a sequence within the center region of human DHX16. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (100%), Monkey (99%), Pig (100%), Rat (100%), Zebrafish (88%), Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction