missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHX16 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA530272

Product Code. 13434767

  • € 526.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (88%), Pig (100%), Rhesus Monkey (99%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a functional homolog of fission yeast Prp8 protein involved in cell cycle progression. This gene is mapped to the MHC region on chromosome 6p21.3, a region where many malignant, genetic and autoimmune disease genes are linked. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DHX16
Polyclonal
Unconjugated
Dhx16
2410006N22Rik; ATP-dependent RNA helicase #3; DBP2; Ddx16; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 16; DEAD/H box 16; DEAH (Asp-Glu-Ala-His) box polypeptide 16; DEAH-box helicase 16; DEAH-box protein 16; Dhx16; KIAA0577; mKIAA0577; Pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16; pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16; PRO2014; Prp2; PRP8; PRPF2; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16; RNA helicase
Rabbit
Antigen affinity chromatography
RUO
8449
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.52 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
O60231
Dhx16
Recombinant protein encompassing a sequence within the center region of human DHX16. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.