missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX35 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | DHX35 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18482292
|
Novus Biologicals
NBP2-30588-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18178412
|
Novus Biologicals
NBP2-30588 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHX35 Polyclonal specifically detects DHX35 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DHX35 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C20orf15, DDX35, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 35, DEAH (Asp-Glu-Ala-His) box polypeptide 35, DEAH box protein 35, DEAH-box protein 35, EC 3.6.1, EC 3.6.4.13, FLJ22759, KAIA0875, probable ATP-dependent RNA helicase DHX35 | |
| DHX35 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9H5Z1 | |
| 60625 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SQEILSIAAMMQIQNIFVVPPNQKSHAIRVHRKFAVEEGDHLTMLNIYEAFIKHNKDSKWCQEHFLNYKGLVRAATVREQLKKLLVKFQVPRKSSEGDP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title