missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX35 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 590.10
Specifications
| Antigen | DHX35 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18205504
|
Novus Biologicals
NBP2-57313 |
100 μL |
€ 624.00 € 590.10 / 100µL Save € 33.90 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628347
|
Novus Biologicals
NBP2-57313-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHX35 Polyclonal specifically detects DHX35 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| DHX35 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C20orf15, DDX35, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 35, DEAH (Asp-Glu-Ala-His) box polypeptide 35, DEAH box protein 35, DEAH-box protein 35, EC 3.6.1, EC 3.6.4.13, FLJ22759, KAIA0875, probable ATP-dependent RNA helicase DHX35 | |
| DHX35 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 60625 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VLRCIVSGFFANAARFHSTGAYRTIRDDHELHIHPASVLYAEKPPRWVIYNEVIQTSKYYMRDVTAIESAWLLELAPHFYQQGTHLSLKAKRAKVQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title