missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHX36 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16387334
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16387334 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16387334 Supplier Invitrogen™ Supplier No. PA578644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2 Predicted reactivity: Mouse (82%), Rat (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure. DHX36 (DEAH box protein 36), also known as MLE-like protein 1 and RNA helicase associated with AU-rich element ARE (RHAU), belongs to RNA helicase of the DEAH family and may function in sex development and spermatogenesis. It is expressed in testis and is evolutionary conserved with true orthologs in almost all animal species. DHX36 plays a role in degradation and deadenylation of mRNAs containing in their 3'-UTR the consensus ARE sequence element. DHX36 is required for early embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX36
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene DHX36
Gene Accession No. Q8VHK9, Q9H2U1
Gene Alias 2810407E23Rik; AI452301; Asp-Glu-Ala-His; ATP-dependent DNA/RNA helicase DHX36; ATP-dependent RNA helicase DHX36; AU022184; Ddx36; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36; DEAD/H box polypeptide 36; DEAH (Asp-Glu-Ala-His) box polypeptide 36; DEAH box protein 36; DEAH-box helicase 36; DEAH-box protein 36; DHX36; G4 resolvase-1; G4R1; G4-resolvase 1; G4-resolvase-1; KIAA1488; mKIAA1488; Mlel1; MLE-like protein 1; probable ATP-dependent RNA helicase DHX36; RHAU; RNA helicase associated with AU-rich element ARE; RNA helicase associated with AU-rich element protein
Gene Symbols DHX36
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human DHX36. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170506, 310461, 72162
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.