missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX58 Antibody (6A6A3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61751
This item is not returnable.
View return policy
Description
DHX58 Monoclonal specifically detects DHX58 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| DHX58 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| D11lgp2e, D11LGP2probable ATP-dependent RNA helicase DHX58, DEXH (Asp-Glu-X-His) box polypeptide 58, EC 3.6.4.13, LGP2ortholog of mouse D11lgp2, Probable ATP-dependent helicase LGP2, Protein D11Lgp2 homolog, RIG-I-like receptor LGP2, RLR, RNA helicase LGP2 | |
| Mouse | |
| 76.6 kDa | |
| 0.1 mL | |
| 79132 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 6A6A3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| DHX58 | |
| Purified recombinant fragment of human DHX58 (AA: 479-678) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction