missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX58 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76390CL1
This item is not returnable.
View return policy
Description
DHX58 Polyclonal antibody specifically detects DHX58 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DHX58 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| D11lgp2e, D11LGP2probable ATP-dependent RNA helicase DHX58, DEXH (Asp-Glu-X-His) box polypeptide 58, EC 3.6.4.13, LGP2ortholog of mouse D11lgp2, Probable ATP-dependent helicase LGP2, Protein D11Lgp2 homolog, RIG-I-like receptor LGP2, RLR, RNA helicase LGP2 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human LGP2. The immunogen is located within amino acids 90 - 140 of LGP2. Amino Acid Squence: ALTSPEEEEHVELT | |
| 0.1 mL | |
| Innate Immunity | |
| 79132 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction