missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-2-thienylglycolic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-2,2-dithiophen-2-ylacetic Acid |
| CAS | 4746-63-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H8 O3 S2 |
| Quantity | 20 mg |
| Synonym | Glycolic acid, di-2-thienyl- (7CI,8CI), (Hydroxy)(di-2-thienyl)acetic acid, 2-Hydroxy-2,2-di(2-thienyl)acetic acid, Di-2-thienylglycolic acid, alpha,alpha-Di(2-thienyl)glycolic acid, Tiotropium Bromide Monohydrate Imp A (EP) |
| InChI Formula | InChI=1S/C10H8O3S2/c11-9(12)10(13,7-3-1-5-14-7)8-4-2-6-15-8/h1-6,13H,(H,11,12) |
| SMILES | OC(=O)C(O)(c1cccs1)c2cccs2 |
| IUPAC Name | 2-hydroxy-2,2-dithiophen-2-ylacetic acid |
| Molecular Weight (g/mol) | 240.3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?