Learn More
Di-iso-propyl-d14 Ether, CDN
Product Code. 30313665
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313665 0.05 g 0.05g
30309146 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30313665 Supplier CDN Supplier No. CDND80500.05G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diisopropyl Ether-D14
CAS 201489-40-9
Molecular Formula C6 D14 O
Quantity 0.05 g
Synonym Propane-1,1,1,2,3,3,3-d7, 2,2'-oxybis- (9 CI), 2,2'-Oxybis[propane-1,1,1,2,3,3,3-d7], Isopropyl ether-d14, Diisopropyl ether-D14, Diisopropyl-d14 ether, Diisopropylether-d14, Di-iso-propyl-d14 Ether
InChI Formula InChI=1 S/C6H14O/c1-5(2)7-6(3)4/h5-6 H,1-4H3/i1D3,2D3,3D3,4D3,5 D,6 D
SMILES [2 H]C([2 H])([2 H])C([2 H])(OC([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC Name 1,1,1,2,3,3,3-heptadeuterio-2-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yloxy)propane
Molecular Weight (g/mol) 116.261
Formula Weight 116.1923 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity