missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-iso-propyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-isopropyl ester D4 |
| CAS | 2708282-44-2 |
| Molecular Formula | C14 2H4 H14 O4 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C14H18O4/c1-9(2)17-13(15)11-7-5-6-8-12(11)14(16)18-10(3)4/h5-10 H,1-4H3/i5D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC(C)C)c(C(=O)OC(C)C)c1[2 H] |
| IUPAC Name | dipropan-2-yl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 254.32 |
| Formula Weight | 254.146 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction