Learn More
Di-tert-butyl Maleate, TRC
Specifications
Specifications
| Chemical Name or Material | Di-tert-butyl Maleate |
| CAS | 18305-60-7 |
| Molecular Formula | C12H20O4 |
| Quantity | 1 g |
| Synonym | (Z)-2-Butenedioic Acid Bis(1,1-dimethylethyl) Ester, Maleic Acid Di-tert-butyl Ester |
| InChI Formula | InChI=1S/C12H20O4/c1-11(2,3)15-9(13)7-8-10(14)16-12(4,5)6/h7-8H,1-6H3/b8-7- |
| SMILES | O=C(OC(C)(C)C)/C=C\C(OC(C)(C)C)=O |
| IUPAC Name | di-tert-butyl maleate |
| Molecular Weight (g/mol) | 228.28 |
| Formula Weight | 228.14 |